Anti-SLC2A1 Mouse Monoclonal Antibody [clone: 202921]

Supplier: R & D Systems
MAB14181
102885-268EA 633.42 USD
102885-268
Anti-SLC2A1 Mouse Monoclonal Antibody [clone: 202921]
Antibodies
Simple Western

Type: Primary
Antigen: SLC2A1
Clonality: Monoclonal
Clone: 202921
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR