Anti-SLC2A1 Mouse Monoclonal Antibody [clone: 202915]

Supplier: R & D Systems
MAB1418
220019-068EA 623.29 USD
220019-068
Anti-SLC2A1 Mouse Monoclonal Antibody [clone: 202915]
Antibodies
Type: Primary
Antigen: SLC2A1
Clonality: Monoclonal
Clone: 202915
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR