Anti-INS (C-Peptide) Mouse Monoclonal Antibody [clone: 790904]

Supplier: R & D Systems
MAB14171
102885-266EA 594.37 USD
102885-266
Anti-INS (C-Peptide) Mouse Monoclonal Antibody [clone: 790904]
Antibodies
Type: Primary
Antigen: INS (C-Peptide)
Clonality: Monoclonal
Clone: 790904
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR