Anti-A2B5 Mouse Monoclonal Antibody [clone: 15]

Supplier: R & D Systems
MAB1416
220019-066EA 647.88 USD
220019-066
Anti-A2B5 Mouse Monoclonal Antibody [clone: 15]
Antibodies
Type: Primary
Antigen: A2B5
Clonality: Monoclonal
Clone: 15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR