Anti-SLC2A3 Mouse Monoclonal Antibody [clone: 202017]

Supplier: R & D Systems
MAB1415
220019-065EA 616.06 USD
220019-065
Anti-SLC2A3 Mouse Monoclonal Antibody [clone: 202017]
Antibodies
Type: Primary
Antigen: SLC2A3
Clonality: Monoclonal
Clone: 202017
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR