Anti-SLC2A2 Mouse Monoclonal Antibody [clone: 199017]

Supplier: R & D Systems
MAB1414
220019-064EA 623.29 USD
220019-064
Anti-SLC2A2 Mouse Monoclonal Antibody [clone: 199017]
Antibodies
Type: Primary
Antigen: SLC2A2
Clonality: Monoclonal
Clone: 199017
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR