Anti-FGFBP1 Mouse Monoclonal Antibody [clone: 227239]

Supplier: R & D Systems
MAB1413
220019-063EA 763.57 USD
220019-063
Anti-FGFBP1 Mouse Monoclonal Antibody [clone: 227239]
Antibodies
Type: Primary
Antigen Symbol: FGFBP1
Antigen Name: fibroblast growth factor binding protein 1
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 227239
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR