Anti-CFC1 Mouse Monoclonal Antibody [clone: 219708]

Supplier: R & D Systems
MAB1410
220019-062EA 772.25 USD
220019-062
Anti-CFC1 Mouse Monoclonal Antibody [clone: 219708]
Antibodies
Type: Primary
Antigen: CFC1
Clonality: Monoclonal
Clone: 219708
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR