Anti-CSTB Rat Monoclonal Antibody [clone: 227807]

Supplier: R & D Systems
MAB14091
220019-060EA 763.57 USD
220019-060
Anti-CSTB Rat Monoclonal Antibody [clone: 227807]
Antibodies
Type: Primary
Antigen: CSTB
Clonality: Monoclonal
Clone: 227807
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR