Anti-CSTB Mouse Monoclonal Antibody [clone: 225228]

Supplier: R & D Systems
MAB1408
220019-058EA 763.57 USD
220019-058
Anti-CSTB Mouse Monoclonal Antibody [clone: 225228]
Antibodies
Type: Primary
Antigen: CSTB
Clonality: Monoclonal
Clone: 225228
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR