Anti-CSTA Mouse Monoclonal Antibody [clone: 224705]

Supplier: R & D Systems
MAB1407
220019-057EA 772.25 USD
220019-057
Anti-CSTA Mouse Monoclonal Antibody [clone: 224705]
Antibodies
Type: Primary
Antigen: CSTA
Clonality: Monoclonal
Clone: 224705
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR