Anti-IL23R Mouse Monoclonal Antibody [clone: 218213]

Supplier: R & D Systems
MAB14001
220019-053EA 639.2 USD
220019-053
Anti-IL23R Mouse Monoclonal Antibody [clone: 218213]
Antibodies
Type: Primary
Antigen: IL23R
Clonality: Monoclonal
Clone: 218213
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR