Anti-IL25 Rat Monoclonal Antibody [clone: 881014]

Supplier: R & D Systems
MAB13992
102885-264EA 633.42 USD
102885-264
Anti-IL25 Rat Monoclonal Antibody [clone: 881014]
Antibodies
Type: Primary
Antigen: IL25
Clonality: Monoclonal
Clone: 881014
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR