Anti-TSLP Mouse Monoclonal Antibody [clone: 258136]

Supplier: R & D Systems
MAB1398
220019-049EA 647.88 USD
220019-049
Anti-TSLP Mouse Monoclonal Antibody [clone: 258136]
Antibodies
Type: Primary
Antigen: TSLP
Clonality: Monoclonal
Clone: 258136
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR