Anti-IGDCC4 Rat Monoclonal Antibody [clone: 261123]

Supplier: R & D Systems
MAB1394
220019-048EA 561.11 USD
220019-048
Anti-IGDCC4 Rat Monoclonal Antibody [clone: 261123]
Antibodies
Type: Primary
Antigen: IGDCC4
Clonality: Monoclonal
Clone: 261123
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR