Anti-DLL4 Rat Monoclonal Antibody [clone: 207822]

Supplier: R & D Systems
MAB1389
220019-045EA 779.48 USD
220019-045
Anti-DLL4 Rat Monoclonal Antibody [clone: 207822]
Antibodies
Type: Primary
Antigen: DLL4
Clonality: Monoclonal
Clone: 207822
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR