Anti-CDH2 Mouse Monoclonal Antibody [clone: 691701]

Supplier: R & D Systems
MAB13882
102885-262EA 594.37 USD
102885-262
Anti-CDH2 Mouse Monoclonal Antibody [clone: 691701]
Antibodies
Type: Primary
Antigen: CDH2
Clonality: Monoclonal
Clone: 691701
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR