Anti-JNK1 Mouse Monoclonal Antibody [clone: 252355]

Supplier: R & D Systems
MAB1387
220019-044EA 594.37 USD
220019-044
Anti-JNK1 Mouse Monoclonal Antibody [clone: 252355]
Antibodies
Type: Primary
Antigen: JNK1
Clonality: Monoclonal
Clone: 252355
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR