Anti-RELT Mouse Monoclonal Antibody [clone: 238104]

Supplier: R & D Systems
MAB1385
220019-043EA 763.57 USD
220019-043
Anti-RELT Mouse Monoclonal Antibody [clone: 238104]
Antibodies
Type: Primary
Antigen: RELT
Clonality: Monoclonal
Clone: 238104
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR