Anti-KLRC2 Mouse Monoclonal Antibody [clone: 134522]

Supplier: R & D Systems
MAB1381-100
102885-258EA 655.11 USD
102885-258
Anti-KLRC2 Mouse Monoclonal Antibody [clone: 134522]
Antibodies
Type: Primary
Antigen: KLRC2
Clonality: Monoclonal
Clone: 134522
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR