Anti-KLRC2 Mouse Monoclonal Antibody [clone: 134591]

Supplier: R & D Systems
MAB138-100
102885-256EA 655.11 USD
102885-256
Anti-KLRC2 Mouse Monoclonal Antibody [clone: 134591]
Antibodies
Type: Primary
Antigen: KLRC2
Clonality: Monoclonal
Clone: 134591
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR