Anti-REG4 Mouse Monoclonal Antibody [clone: 200214]

Supplier: R & D Systems
MAB1379
220019-039EA 772.25 USD
220019-039
Anti-REG4 Mouse Monoclonal Antibody [clone: 200214]
Antibodies
Type: Primary
Antigen: REG4
Clonality: Monoclonal
Clone: 200214
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR