Anti-GZMH Mouse Monoclonal Antibody [clone: 185813]

Supplier: R & D Systems
MAB1377
220019-038EA 763.57 USD
220019-038
Anti-GZMH Mouse Monoclonal Antibody [clone: 185813]
Antibodies
Type: Primary
Antigen Symbol: GZMH
Antigen Name: granzyme H (cathepsin G-like 2, protein h-CCPX)
Antigen Synonyms: CCP-X, CGL-2, CSP-C, CTLA1, CTSGL2
Clonality: Monoclonal
Clone: 185813
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR