Anti-LTA Mouse Monoclonal Antibody [clone: 135125]

Supplier: R & D Systems
MAB1370
220019-035EA 763.57 USD
220019-035
Anti-LTA Mouse Monoclonal Antibody [clone: 135125]
Antibodies
Type: Primary
Antigen: LTA
Clonality: Monoclonal
Clone: 135125
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR