Anti-AFP Mouse Monoclonal Antibody [clone: 21417]

Supplier: R & D Systems
MAB13691
220019-034EA 763.57 USD
220019-034
Anti-AFP Mouse Monoclonal Antibody [clone: 21417]
Antibodies
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: 21417
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR