Anti-AFP Mouse Monoclonal Antibody [clone: 18952]

Supplier: R & D Systems
MAB1368
220019-032EA 647.88 USD
220019-032
Anti-AFP Mouse Monoclonal Antibody [clone: 18952]
Antibodies
Simple Western

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: 18952
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR