Anti-ANGPTL3 Rat Monoclonal Antibody [clone: 128611]

Supplier: R & D Systems
MAB13621
102885-254EA 772.25 USD
102885-254
Anti-ANGPTL3 Rat Monoclonal Antibody [clone: 128611]
Antibodies
Type: Primary
Antigen: ANGPTL3
Clonality: Monoclonal
Clone: 128611
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR