Anti-ANGPTL3 Rat Monoclonal Antibody [clone: 128644]

Supplier: R & D Systems
MAB1362
220019-029EA 577.02 USD
220019-029
Anti-ANGPTL3 Rat Monoclonal Antibody [clone: 128644]
Antibodies
Type: Primary
Antigen: ANGPTL3
Clonality: Monoclonal
Clone: 128644
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR