Anti-TP53 Mouse Monoclonal Antibody [clone: 184727]

Supplier: R & D Systems
MAB13551
220019-023EA 594.37 USD
220019-023
Anti-TP53 Mouse Monoclonal Antibody [clone: 184727]
Antibodies
Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: 184727
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR