Anti-TRP53 Mouse Monoclonal Antibody [clone: 184721]

Supplier: R & D Systems
MAB1355
220019-021EA 655.11 USD
220019-021
Anti-TRP53 Mouse Monoclonal Antibody [clone: 184721]
Antibodies
Type: Primary
Antigen: TRP53
Clonality: Monoclonal
Clone: 184721
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR