Anti-ITGA6 Rat Monoclonal Antibody [clone: GoH3]

Supplier: R & D Systems
MAB13501
220019-018EA 655.11 USD
220019-018
Anti-ITGA6 Rat Monoclonal Antibody [clone: GoH3]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: ITGA6
Clonality: Monoclonal
Clone: GoH3
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR