Anti-ITGA6 Mouse Monoclonal Antibody [clone: MP4F10]

Supplier: R & D Systems
MAB1350
220019-017EA 763.57 USD
220019-017
Anti-ITGA6 Mouse Monoclonal Antibody [clone: MP4F10]
Antibodies
Type: Primary
Antigen: ITGA6
Clonality: Monoclonal
Clone: MP4F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR