Anti-SLC2A5 Mouse Monoclonal Antibody [clone: 195205]

Supplier: R & D Systems
MAB1349
220019-016EA 608.83 USD
220019-016
Anti-SLC2A5 Mouse Monoclonal Antibody [clone: 195205]
Antibodies
Type: Primary
Antigen: SLC2A5
Clonality: Monoclonal
Clone: 195205
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR