Anti-PPP3R1 Mouse Monoclonal Antibody [clone: 212306]

Supplier: R & D Systems
MAB1348
220019-015EA 594.37 USD
220019-015
Anti-PPP3R1 Mouse Monoclonal Antibody [clone: 212306]
Antibodies
Type: Primary
Antigen: PPP3R1
Clonality: Monoclonal
Clone: 212306
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR