Anti-MAPK12 Mouse Monoclonal Antibody [clone: 212464]

Supplier: R & D Systems
MAB1347
220019-014EA 594.37 USD
220019-014
Anti-MAPK12 Mouse Monoclonal Antibody [clone: 212464]
Antibodies
Simple Western

Type: Primary
Antigen: MAPK12
Clonality: Monoclonal
Clone: 212464
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR