Anti-GZMG Rat Monoclonal Antibody [clone: 207639]

Supplier: R & D Systems
MAB1346
220019-013EA 763.57 USD
220019-013
Anti-GZMG Rat Monoclonal Antibody [clone: 207639]
Antibodies
Type: Primary
Antigen Symbol: GZMG
Antigen Name: granzyme G
Antigen Synonyms: Ctla-7, Ctla7
Clonality: Monoclonal
Clone: 207639
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR