Anti-ITGA3 Mouse Monoclonal Antibody [clone: IA3]

Supplier: R & D Systems
MAB1345
220019-011EA 772.25 USD
220019-011
Anti-ITGA3 Mouse Monoclonal Antibody [clone: IA3]
Antibodies
Type: Primary
Antigen: ITGA3
Clonality: Monoclonal
Clone: IA3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR