Anti-WIF1 Mouse Monoclonal Antibody [clone: 133009]

Supplier: R & D Systems
MAB1342
220019-007EA 763.57 USD
220019-007
Anti-WIF1 Mouse Monoclonal Antibody [clone: 133009]
Antibodies
Type: Primary
Antigen: WIF1
Clonality: Monoclonal
Clone: 133009
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR