Anti-CD86 Mouse Monoclonal Antibody [clone: 199622]

Supplier: R & D Systems
MAB13401
220019-005EA 763.57 USD
220019-005
Anti-CD86 Mouse Monoclonal Antibody [clone: 199622]
Antibodies
Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: 199622
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR