Anti-CD86 Mouse Monoclonal Antibody [clone: 199602]

Supplier: R & D Systems
MAB1340
220019-004EA 763.57 USD
220019-004
Anti-CD86 Mouse Monoclonal Antibody [clone: 199602]
Antibodies
Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: 199602
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR