Anti-WIF1 Mouse Monoclonal Antibody [clone: 133015]

Supplier: R & D Systems
MAB134-500 MAB134-050
220019-012EA 815.63 USD
220019-012 220019-006
Anti-WIF1 Mouse Monoclonal Antibody [clone: 133015]
Antibodies
Type: Primary
Antigen: WIF1
Clonality: Monoclonal
Clone: 133015
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR