Anti-LGALS7 Mouse Monoclonal Antibody [clone: 198614]

Supplier: R & D Systems
MAB1339
220019-003EA 763.57 USD
220019-003
Anti-LGALS7 Mouse Monoclonal Antibody [clone: 198614]
Antibodies
Type: Primary
Antigen: LGALS7
Clonality: Monoclonal
Clone: 198614
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR