Anti-SNCA Mouse Monoclonal Antibody [clone: 189105]

Supplier: R & D Systems
MAB1338
220019-002EA 779.48 USD
220019-002
Anti-SNCA Mouse Monoclonal Antibody [clone: 189105]
Antibodies
Type: Primary
Antigen: SNCA
Clonality: Monoclonal
Clone: 189105
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR