Anti-INS Mouse Monoclonal Antibody [clone: 253627]

Supplier: R & D Systems
MAB13361
220019-001EA 655.11 USD
220019-001
Anti-INS Mouse Monoclonal Antibody [clone: 253627]
Antibodies
Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: 253627
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR