Anti-IL17F Mouse Monoclonal Antibody [clone: 775620]

Supplier: R & D Systems
MAB13352
102885-250EA 772.25 USD
102885-250
Anti-IL17F Mouse Monoclonal Antibody [clone: 775620]
Antibodies
Type: Primary
Antigen: IL17F
Clonality: Monoclonal
Clone: 775620
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR