Anti-IL17F Mouse Monoclonal Antibody [clone: 197301]

Supplier: R & D Systems
MAB13351
220019-000EA 594.37 USD
220019-000
Anti-IL17F Mouse Monoclonal Antibody [clone: 197301]
Antibodies
Type: Primary
Antigen: IL17F
Clonality: Monoclonal
Clone: 197301
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR