Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 196618]

Supplier: R & D Systems
MAB13291-050 MAB13291-100
220018-995EA 633.42 USD
220018-995 220018-994
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 196618]
Antibodies
Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone: 196618
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR