Anti-O1 Mouse Monoclonal Antibody [clone: O1]

Supplier: R & D Systems
MAB1327
220018-992EA 475.78 USD
220018-992
Anti-O1 Mouse Monoclonal Antibody [clone: O1]
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: Oligodendrocyte Marker O1
Antigen Synonyms:
Clonality: Monoclonal
Clone: O1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR