Anti-O4 Mouse Monoclonal Antibody [clone: O4]

Supplier: R & D Systems
MAB1326
220018-991EA 475.78 USD
220018-991
Anti-O4 Mouse Monoclonal Antibody [clone: O4]
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: Oligodendrocyte Marker O4
Antigen Synonyms:
Clonality: Monoclonal
Clone: O4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR