Anti-ENG Rat Monoclonal Antibody [clone: 209701]

Supplier: R & D Systems
MAB1320
220018-987EA 647.88 USD
220018-987
Anti-ENG Rat Monoclonal Antibody [clone: 209701]
Antibodies
Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone: 209701
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR